4-Pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: Pyridine-4-boronic acid pinacol ester, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

PN-2352-5G PN-2352-25G PN-2352-1G
COBBPN-2352-5GEA 111 EUR
COBBPN-2352-5G COBBPN-2352-25G COBBPN-2352-1G
4-Pyridinboronsäurepinakolester
4-Pyridinboronsäurepinakolester
Formel: C₁₁H₁₆BNO₂
Molecular Weight: 205,06 g/mol
MDL: MFCD01319051
CAS-Nummer: 181219-01-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR