3-Pyridinboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
Pyridine-3-boronic acid pinacol ester, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
PN-2359-5G
PN-2359-1G
COBBPN-2359-5GEA
50.9
EUR
COBBPN-2359-5G
COBBPN-2359-1G
3-Pyridinboronsäurepinakolester
3-Pyridinboronsäurepinakolester
Formel:
C₁₁H₁₆BNO₂ Molecular Weight: 205.06 g/mol |
MDL:
MFCD03084758 CAS-Nummer: 329214-79-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...