2-Chlor-5-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-(6-Chloro-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Chloropyridine-5-boronic acid Pinacol ester, 2-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

PN-2901-5G PN-2901-25G PN-2901-1G
COBBPN-2901-5GEA 77.3 EUR
COBBPN-2901-5G COBBPN-2901-25G COBBPN-2901-1G
2-Chlor-5-pyridinboronsäurepinakolester
2-Chlor-5-pyridinboronsäurepinakolester
Formel: C₁₁H₁₅BClNO₂
Molecular Weight: 239.51 g/mol
MDL: MFCD03412793
CAS-Nummer: 444120-94-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR