2-Brom-5-pyridinboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
2-Bromopyridine-5-boronic acid pinacol ester, 2-Bromo-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
PN-3423-1G
PN-3423-5G
COBBPN-3423-1GEA
73.7
EUR
COBBPN-3423-1G
COBBPN-3423-5G
2-Brom-5-pyridinboronsäurepinakolester
2-Brom-5-pyridinboronsäurepinakolester
Formel:
C₁₁H₁₅BBrNO₂ Molecular Weight: 283.96 g/mol |
MDL:
MFCD03412794 CAS-Nummer: 214360-62-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...