3-Chlor-4-pyridinboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
3-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 3-Chloropyridine-4-boronic acid pinacol ester
PN-3442-5G
PN-3442-1G
COBBPN-3442-5GEA
958
EUR
COBBPN-3442-5G
COBBPN-3442-1G
3-Chlor-4-pyridinboronsäurepinakolester
3-Chlor-4-pyridinboronsäurepinakolester
Formel:
C₁₁H₁₅BClNO₂ Molecular Weight: 239,51 g/mol |
MDL:
MFCD06798249 CAS-Nummer: 458532-90-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...