2-Fluor-5-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-(6-Fluoro-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 6-Fluoropyridine-3-boronic acid Pinacol ester, 2-Fluoropyridine-5-boronic acid pinacol ester

PN-3448-1G PN-3448-5G
COBBPN-3448-1GEA 22 EUR
COBBPN-3448-1G COBBPN-3448-5G
2-Fluor-5-pyridinboronsäurepinakolester
2-Fluor-5-pyridinboronsäurepinakolester
Formel: C₁₁H₁₅BFNO₂
Molecular Weight: 223,05 g/mol
MDL: MFCD03412792
CAS-Nummer: 444120-95-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR