4-Chlor-3-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 4-Chloropyridine-3-boronic Acid Pinacol Ester

PN-3449-1G PN-3449-5G
COBBPN-3449-1GEA 324 EUR
COBBPN-3449-1G COBBPN-3449-5G
4-Chlor-3-pyridinboronsäurepinakolester
4-Chlor-3-pyridinboronsäurepinakolester
Formel: C₁₁H₁₅BClNO₂
Molecular Weight: 239,51 g/mol
MDL: MFCD18914522
CAS-Nummer: 452972-15-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR