2-Fluor-3-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-Fluoropyridine-3-boronic acid pinacol ester, 2-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

PN-3476-5G PN-3476-1G
COBBPN-3476-5GEA 193 EUR
COBBPN-3476-5G COBBPN-3476-1G
2-Fluor-3-pyridinboronsäurepinakolester
2-Fluor-3-pyridinboronsäurepinakolester
Formel: C₁₁H₁₅BFNO₂
Molecular Weight: 223,05 g/mol
MDL: MFCD06798245
CAS-Nummer: 452972-14-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR