2,3-Dichlor-4-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2,3-Dichloropyridine-4-boronic acid, pinacol ester, 2,3-Dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

PN-3926-5G PN-3926-1G
COBBPN-3926-5GEA 273 EUR
COBBPN-3926-5G COBBPN-3926-1G
2,3-Dichlor-4-pyridinboronsäurepinakolester
2,3-Dichlor-4-pyridinboronsäurepinakolester
Formel: C₁₁H₁₄BCl₂NO₂
Molecular Weight: 273.95 g/mol
MDL: MFCD06798257
CAS-Nummer: 1073353-78-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR