2-Hydroxy-5-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-ol, 6-Hydroxypyridine-3-boronic acid pinacol ester

PN-5130-1G PN-5130-5G
COBBPN-5130-1GEA 127 EUR
COBBPN-5130-1G COBBPN-5130-5G
2-Hydroxy-5-pyridinboronsäurepinakolester
2-Hydroxy-5-pyridinboronsäurepinakolester
Formel: C₁₁H₁₆BNO₃
Molecular Weight: 221,06 g/mol
MDL: MFCD06795671
CAS-Nummer: 1054483-78-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR