5-Cyan-2-pyridinboronsäurepinakolester
Lieferant: COMBI-BLOCKS
PN-5145-1G
PN-5145-250M
COBBPN-5145-1GEA
0
EUR
COBBPN-5145-1G
COBBPN-5145-250M
5-Cyan-2-pyridinboronsäurepinakolester
5-Cyan-2-pyridinboronsäurepinakolester
Formel:
C₁₂H₁₅BN₂O₂ |
MDL:
MFCD07367521 CAS-Nummer: 1073353-83-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...