2-Isopropoxy-3-pyridinboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
2-Isopropoxy-3-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-pyridine, 2-Isopropoxypyridine-3-boronic acid pinacol ester, 2-Isopropoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
PN-5202-5G
PN-5202-1G
COBBPN-5202-5GEA
886
EUR
COBBPN-5202-5G
COBBPN-5202-1G
2-Isopropoxy-3-pyridinboronsäurepinakolester
2-Isopropoxy-3-pyridinboronsäurepinakolester
Formel:
C₁₄H₂₂BNO₃ Molecular Weight: 263.14 g/mol |
MDL:
MFCD07366722 CAS-Nummer: 848243-25-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...