2-Chlor-6-fluor-4-pyridinboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
2-Chloro-6-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
PN-5459-1G
PN-5459-250M
COBBPN-5459-1GEA
1180
EUR
COBBPN-5459-1G
COBBPN-5459-250M
2-Chlor-6-fluor-4-pyridinboronsäurepinakolester
2-Chlor-6-fluor-4-pyridinboronsäurepinakolester
Formel:
C₁₁H₁₄BClFNO₂ Molecular Weight: 257.5 g/mol |
MDL:
MFCD11504972 CAS-Nummer: 1146615-89-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...