2,6-Dimethyl-4-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2,6-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

PN-5673-1G PN-5673-250M
COBBPN-5673-1GEA 701 EUR
COBBPN-5673-1G COBBPN-5673-250M
2,6-Dimethyl-4-pyridinboronsäurepinakolester
2,6-Dimethyl-4-pyridinboronsäurepinakolester
Formel: C₁₃H₂₀BNO₂
Molecular Weight: 233.12 g/mol
MDL: MFCD11617853
CAS-Nummer: 325142-95-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR