2-Amino-3-nitro-5-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-5990-5G PN-5990-1G PN-5990-250M
COBBPN-5990-5GEA 1810 EUR
COBBPN-5990-5G COBBPN-5990-1G COBBPN-5990-250M
2-Amino-3-nitro-5-pyridinboronsäurepinakolester
2-Amino-3-nitro-5-pyridinboronsäurepinakolester
Formel: C₁₁H₁₆BN₃O₄
Molecular Weight: 265,08 g/mol
MDL: MFCD11867866
CAS-Nummer: 1032758-80-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR