2,6-Difluor-4-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2,6-Difluoropyridine-4-boronic acid pinacol ester, 2,6-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

PN-8468-1G PN-8468-250M
COBBPN-8468-1GEA 595 EUR
COBBPN-8468-1G COBBPN-8468-250M
2,6-Difluor-4-pyridinboronsäurepinakolester
2,6-Difluor-4-pyridinboronsäurepinakolester
Formel: C₁₁H₁₄BF₂NO₂
Molecular Weight: 241,04 g/mol
MDL: MFCD12913987
CAS-Nummer: 1204333-58-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR