2,6-Dimethoxy-3-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-8610-5G PN-8610-1G
COBBPN-8610-5GEA 715 EUR
COBBPN-8610-5G COBBPN-8610-1G
2,6-Dimethoxy-3-pyridinboronsäurepinakolester
2,6-Dimethoxy-3-pyridinboronsäurepinakolester
Formel: C₁₃H₂₀BNO₄
Molecular Weight: 265.12 g/mol
MDL: MFCD03788240
CAS-Nummer: 214360-59-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR