2-Amino-5-pyridinboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
6-Aminopyridine-3-boronic acid pinacol ester, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine, 2-Aminopyridine-5-boronic acid pinacol ester
PN-8636-5G
PN-8636-1G
COBBPN-8636-5GEA
334
EUR
COBBPN-8636-5G
COBBPN-8636-1G
2-Amino-5-pyridinboronsäurepinakolester
2-Amino-5-pyridinboronsäurepinakolester
Formel:
C₁₁H₁₇BN₂O₂ Molecular Weight: 220,08 g/mol |
MDL:
MFCD05663837 CAS-Nummer: 827614-64-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...