2-Cyan-3-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-Cyanopyridine-3-boronic acid, pinacol ester, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)picolinonitrile

PN-8744-250M PN-8744-100M
COBBPN-8744-250MEA 398 EUR
COBBPN-8744-250M COBBPN-8744-100M
2-Cyan-3-pyridinboronsäurepinakolester
2-Cyan-3-pyridinboronsäurepinakolester
Formel: C₁₂H₁₅BN₂O₂
Molecular Weight: 230.07 g/mol
MDL: MFCD08458477
CAS-Nummer: 878194-93-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR