4-Cyan-3-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isonicotinonitrile, 4-Cyanopyridine-3-boronic acid, pinacol ester, 4-Cyanopyridine-3-boronic acid pinacol ester

PN-8745-250M PN-8745-1G
COBBPN-8745-250MEA 240 EUR
COBBPN-8745-250M COBBPN-8745-1G
4-Cyan-3-pyridinboronsäurepinakolester
4-Cyan-3-pyridinboronsäurepinakolester
Formel: C₁₂H₁₅BN₂O₂
Molecular Weight: 230.07 g/mol
MDL: MFCD08458478
CAS-Nummer: 878194-91-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR