2-Acetamido-5-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-8757-1G PN-8757-250M
COBBPN-8757-1GEA 0 EUR
COBBPN-8757-1G COBBPN-8757-250M
2-Acetamido-5-pyridinboronsäurepinakolester
2-Acetamido-5-pyridinboronsäurepinakolester
Formel: C₁₃H₁₉BN₂O₃
MDL: MFCD08690229
CAS-Nummer: 904326-87-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR