2-Methoxy-3-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-Methoxypyridine-3-boronic acid pinacol ester, 2-Methoxy-3-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-pyridine, 2-Methoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

PN-8856-5G PN-8856-25G PN-8856-1G
COBBPN-8856-5GEA 227 EUR
COBBPN-8856-5G COBBPN-8856-25G COBBPN-8856-1G
2-Methoxy-3-pyridinboronsäurepinakolester
2-Methoxy-3-pyridinboronsäurepinakolester
Formel: C₁₂H₁₈BNO₃
Molecular Weight: 235,09 g/mol
MDL: MFCD05663507
CAS-Nummer: 532391-31-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR