2-Cyan-5-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-Cyano-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)picolinonitrile, 2-(6-Cyano-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 6-Cyano-3-pyridylboronic acid Pinacol ester, 2-Cyanopyridine-5-boronic acid pinacol ester

PN-8873-5G PN-8873-1G
COBBPN-8873-5GEA 273 EUR
COBBPN-8873-5G COBBPN-8873-1G
2-Cyan-5-pyridinboronsäurepinakolester
2-Cyan-5-pyridinboronsäurepinakolester
Formel: C₁₂H₁₅BN₂O₂
Molecular Weight: 230.07 g/mol
MDL: MFCD06657825
CAS-Nummer: 741709-63-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR