6-Brom-2-pyridinboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
2-Bromo-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 6-Bromo-2-pyridylboronic acid Pinacol ester, 6-Bromopyridine-2-boronic acid pinacol ester, 2-(6-Bromo-2-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PN-8882-5G
PN-8882-1G
COBBPN-8882-5GEA
249
EUR
COBBPN-8882-5G
COBBPN-8882-1G
6-Brom-2-pyridinboronsäurepinakolester
6-Brom-2-pyridinboronsäurepinakolester
Formel:
C₁₁H₁₅BBrNO₂ Molecular Weight: 283.96 g/mol |
MDL:
MFCD06796177 CAS-Nummer: 651358-83-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...