6-Chlor-2-pyridinboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
2-Chloro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 6-Chloropyridine-2-boronic acid pinacol ester
PN-8885-1G
PN-8885-5G
COBBPN-8885-1GEA
188
EUR
COBBPN-8885-1G
COBBPN-8885-5G
6-Chlor-2-pyridinboronsäurepinakolester
6-Chlor-2-pyridinboronsäurepinakolester
Formel:
C₁₁H₁₅BClNO₂ Molecular Weight: 239,51 g/mol |
MDL:
MFCD06798259 CAS-Nummer: 652148-92-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...