6-Methoxy-2-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-Methoxy-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 6-Methoxypyridine-2-boronic acid pinacol ester

PN-8888-5G PN-8888-1G
COBBPN-8888-5GEA 571 EUR
COBBPN-8888-5G COBBPN-8888-1G
6-Methoxy-2-pyridinboronsäurepinakolester
6-Methoxy-2-pyridinboronsäurepinakolester
Formel: C₁₂H₁₈BNO₃
Molecular Weight: 235.09 g/mol
MDL: MFCD06798266
CAS-Nummer: 1034297-69-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR