5-Cyan-3-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)nicotinonitrile, 3-Cyano-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 3-Cyanopyridine-5-boronic acid Pinacol ester, 3-Cyanopyridine-5-boronic acid pinacol ester, 2-(5-Cyano-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PN-8893-5G PN-8893-1G
COBBPN-8893-5GEA 722 EUR
COBBPN-8893-5G COBBPN-8893-1G
5-Cyan-3-pyridinboronsäurepinakolester
5-Cyan-3-pyridinboronsäurepinakolester
Formel: C₁₂H₁₅BN₂O₂
Molecular Weight: 230.07 g/mol
MDL: MFCD07780755
CAS-Nummer: 402718-29-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR