2-Methyl-2-pentensäure
Lieferant: FLUOROCHEM
002778-25G
002778-100G
FLUO002778-25GEA
94.1
EUR
FLUO002778-25G
FLUO002778-100G
2-Methyl-2-pentensäure
2-Methyl-2-pentenoic acid
Formel:
C₆H₁₀O₂ Molecular Weight: 114.14 g/mol |
MDL:
MFCD00002655 CAS-Nummer: 3142-72-1 EINECS: 221-552-0 UN: 3265 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...