(3-Fluor-4-(trifluormethyl)phenyl)essigsäure

Lieferant: FLUOROCHEM
Achtung

Synonyme: 3-Fluoro-4-(trifluoromethyl)phenylacetic acid

007939-1G 007939-5G
FLUO007939-1GEA 55.4 EUR
FLUO007939-1G FLUO007939-5G
(3-Fluor-4-(trifluormethyl)phenyl)essigsäure
(3-Fluor-4-(trifluormethyl)phenyl)essigsäure
Formel: C₉H₆F₄O₂
Molecular Weight: 222.14 g/mol
MDL: MFCD00236303
CAS-Nummer: 238754-67-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR