4-Cyanphenylboronsäurepinakolester

Lieferant: FLUOROCHEM

Synonyme: 4-Cyanobenzeneboronic acid pinacol ester, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile, 4-Cyanophenylboronic acid, pinacol ester

010831-5G 010831-1G
FLUO010831-5GEA 137 EUR
FLUO010831-5G FLUO010831-1G
4-Cyanphenylboronsäurepinakolester
4-Cyanphenylboronsäurepinakolester
Formel: C₁₃H₁₆BNO₂
Molecular Weight: 229,09 g/mol
MDL: MFCD03093897
CAS-Nummer: 171364-82-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR