4-Hydroxyphenylboronsäurepinakolester

Lieferant: FLUOROCHEM

Synonyme: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol, 4-Hydroxybenzeneboronic acid pinacol ester

010837-5G FL10837.20 010837-25G 010837-1G
FLUO010837-5GEA 107 EUR
FLUO010837-5G CHMPFL10837.20 FLUO010837-25G FLUO010837-1G
4-Hydroxyphenylboronsäurepinakolester
4-Hydroxyphenylboronsäurepinakolester
Formel: C₁₂H₁₇BO₃
Molecular Weight: 220.08 g/mol
MDL: MFCD02093756
CAS-Nummer: 269409-70-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR