3-Pyridinboronsäurepinakolester

Lieferant: FLUOROCHEM

Synonyme: Pyridine-3-boronic acid pinacol ester, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

010846-5G 010846-1G
FLUO010846-5GEA 121 EUR
FLUO010846-5G FLUO010846-1G
3-Pyridinboronsäurepinakolester
3-Pyridinboronsäurepinakolester
Formel: C₁₁H₁₆BNO₂
Molecular Weight: 205.06 g/mol
MDL: MFCD03084758
CAS-Nummer: 329214-79-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR