4-Pyridinboronsäurepinakolester
Lieferant: FLUOROCHEM
Synonyme:
Pyridine-4-boronic acid pinacol ester, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
010847-5G
010847-1G
FLUO010847-5GEA
71.7
EUR
FLUO010847-5G
FLUO010847-1G
4-Pyridinboronsäurepinakolester
4-Pyridinboronsäurepinakolester
Formel:
C₁₁H₁₆BNO₂ Molecular Weight: 205.06 g/mol |
MDL:
MFCD01319051 CAS-Nummer: 181219-01-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...