4-Methyl-2-pentensäure

Lieferant: FLUOROCHEM

011183-50G 011183-1G
FLUO011183-50GEA 233 EUR
FLUO011183-50G FLUO011183-1G
4-Methyl-2-pentensäure
4-Methyl-2-pentenoic acid
Formel: C₆H₁₀O₂
Molecular Weight: 114.14 g/mol
MDL: MFCD00043804
CAS-Nummer: 10321-71-8
EINECS: 233-706-4
UN: 3265
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR