(±)-α-Methoxy-α-(trifluormethyl)phenylessigsäure
Lieferant: CHEMPUR
Synonyme:
2-Methoxy-2-phenyl-3,3,3-trifluoropropanoic acid
FL13162.POR
013162-1G
CHMPFL13162.POREA
0
EUR
CHMPFL13162.POR
FLUO013162-1G
(±)-α-Methoxy-α-(trifluormethyl)phenylessigsäure
(±)-α-Methoxy-α-(trifluormethyl)phenylessigsäure
Formel:
C₁₀H₉F₃O₃ Molecular Weight: 234,17 g/mol |
MDL:
MFCD00066637 CAS-Nummer: 81655-41-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...