(2-Brom-4,5-dimethoxyphenyl)essigsäure

Lieferant: FLUOROCHEM

Synonyme: 2-Bromo-4,5-dimethoxyphenylacetic acid

018270-1G 018270-5G
FLUO018270-1GEA 139 EUR
FLUO018270-1G FLUO018270-5G
(2-Brom-4,5-dimethoxyphenyl)essigsäure
(2-Brom-4,5-dimethoxyphenyl)essigsäure
Formel: C₁₀H₁₁BrO₄
Molecular Weight: 275,1 g/mol
MDL: MFCD00016819
CAS-Nummer: 4697-62-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR