3-Methylphenylboronsäurepinakolester
Lieferant: FLUOROCHEM
Synonyme:
3-Methylbenzeneboronic acid, pinacol ester, 4,4,5,5-Tetramethyl-2-(m-tolyl)-1,3,2-dioxaborolane, 3-Tolylboronic acid pinacol ester, m-Tolylboronic acid Pinacol ester, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)toluene
021848.25G
021848-5G
021848-1G
FLUO021848.25GEA
353
EUR
FLUO021848.25G
FLUO021848-5G
FLUO021848-1G
3-Methylphenylboronsäurepinakolester
3-Methylphenylboronsäurepinakolester
Formel:
C₁₃H₁₉BO₂ Molecular Weight: 218.1 g/mol |
MDL:
MFCD05864303 CAS-Nummer: 253342-48-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...