(3-Fluor-4-methoxyphenyl)essigsäure
Lieferant: FLUOROCHEM
Synonyme:
3-Fluoro-4-methoxyphenylacetic acid
021901-5G
021901-1G
FLUO021901-5GEA
41.7
EUR
FLUO021901-5G
FLUO021901-1G
(3-Fluor-4-methoxyphenyl)essigsäure
(3-Fluor-4-methoxyphenyl)essigsäure
Formel:
C₉H₉FO₃ Molecular Weight: 184.17 g/mol |
MDL:
MFCD00060353 CAS-Nummer: 452-14-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...