2-Fluor-5-nitrophenylboronsäure
Lieferant: FLUOROCHEM
Synonyme:
2-Fluoro-5-nitrobenzeneboronic acid
016221-1G
016221-5G
FLUO016221-1GEA
39.6
EUR
FLUO016221-1G
FLUO016221-5G
2-Fluor-5-nitrophenylboronsäure
2-Fluor-5-nitrophenylboronsäure
Formel:
C₆H₅BFNO₄ Molecular Weight: 184.92 g/mol |
MDL:
MFCD03092929 CAS-Nummer: 819849-20-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...