3-Penten-2-on (contains mesityl oxide as impurity), Sigma-Aldrich®

Lieferant: Merck

Synonyme: Pent-3-en-2-one

M-Clarity™ Qualitätsstufe = MQ100

145017-1G 145017-25G 145017-5G
SIAL145017-1GEA 50.4 EUR
SIAL145017-1G SIAL145017-25G SIAL145017-5G
3-Penten-2-on (contains mesityl oxide as impurity), Sigma-Aldrich®
3-Penten-2-on
Formel: C₅H₈O
Molecular Weight: 84,12 g/mol
MDL: MFCD00009290
CAS-Nummer: 625-33-2
EINECS: 210-888-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR