trans-1-Chlor-2-buten, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (E)-1-Chlor-2-buten

M-Clarity™ Qualitätsstufe = MQ100

254584-100G 254584-25G
SIAL254584-100GEA 621 EUR
SIAL254584-100G SIAL254584-25G
trans-1-Chlor-2-buten, Sigma-Aldrich®
trans-1-Chlor-2-buten
Formel: C₄H₇Cl
Molecular Weight: 90,55 g/mol
Lagertemperatur: Kühlschrank
CAS-Nummer: 4894-61-5
EINECS: 225-516-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR