3-Pyridinboronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Pyridine-3-boronic acid pinacol ester, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

M-Clarity™ Qualitätsstufe = MQ100

576565-1G 576565-5G
SIAL576565-1GEA 101 EUR
SIAL576565-1G SIAL576565-5G
3-Pyridinboronsäurepinakolester, Sigma-Aldrich®
3-Pyridinboronsäurepinakolester
Formel: C₁₁H₁₆BNO₂
Molecular Weight: 205,06 g/mol
MDL: MFCD03084758
CAS-Nummer: 329214-79-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR