2-Fluor-5-pyridinboronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-(6-Fluoro-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 6-Fluoropyridine-3-boronic acid Pinacol ester, 2-Fluoropyridine-5-boronic acid pinacol ester

M-Clarity™ Qualitätsstufe = MQ100

592544-1G 592544-5G
SIAL592544-1GEA 64 EUR
SIAL592544-1G SIAL592544-5G
2-Fluor-5-pyridinboronsäurepinakolester, Sigma-Aldrich®
2-Fluor-5-pyridinboronsäurepinakolester
Formel: C₁₁H₁₅BFNO₂
Molecular Weight: 223,05 g/mol
MDL: MFCD03412792
CAS-Nummer: 444120-95-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR