2-Methoxy-5-pyridinboronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 6-Methoxypyridine-3-boronic acid, pinacol ester, 2-Methoxypyridine-5-boronic acid, pinacol ester

M-Clarity™ Qualitätsstufe = MQ200

636029-1G 636029-5G
SIAL636029-1GEA 28.1 EUR
SIAL636029-1G SIAL636029-5G
2-Methoxy-5-pyridinboronsäurepinakolester, Sigma-Aldrich®
2-Methoxy-5-pyridinboronsäurepinakolester
Formel: C₁₂H₁₈BNO₃
Molecular Weight: 235,09 g/mol
MDL: MFCD05663858
CAS-Nummer: 445264-61-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR