2-Amino-5-pyridinboronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 6-Aminopyridine-3-boronic acid pinacol ester, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine, 2-Aminopyridine-5-boronic acid pinacol ester

M-Clarity™ Qualitätsstufe = MQ100

640379-1G 640379-5G
SIAL640379-1GEA 55.1 EUR
SIAL640379-1G SIAL640379-5G
2-Amino-5-pyridinboronsäurepinakolester, Sigma-Aldrich®
2-Amino-5-pyridinboronsäurepinakolester
Formel: C₁₁H₁₇BN₂O₂
Molecular Weight: 220.08 g/mol
MDL: MFCD05663837
CAS-Nummer: 827614-64-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR