4-Nitrophenylboronsäurepinakolester, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
4,4,5,5-Tetramethyl-2-(4-nitrophenyl)-1,3,2-dioxaborolane, 4-Nitrophenylboronic acid Pinacol ester, 1-Nitro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene, 4-Nitrobenzeneboronic acid pinacol ester
M-Clarity™ Qualitätsstufe = MQ100
643890-1G
643890-5G
SIAL643890-1GEA
57.4
EUR
SIAL643890-1G
SIAL643890-5G
4-Nitrophenylboronsäurepinakolester, Sigma-Aldrich®
4-Nitrophenylboronsäurepinakolester
Formel:
C₁₂H₁₆BNO₄ Molecular Weight: 249,07 g/mol |
MDL:
MFCD02179437 CAS-Nummer: 171364-83-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...