(2-Brom-4-methoxyphenyl)essigsäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-(2-Bromo-4-methoxyphenyl)acetic acid, 2-Bromo-4-methoxyphenylacetic acid
M-Clarity™ Qualitätsstufe = MQ100
653195-25G
653195-5G
SIAL653195-25GEA
189
EUR
SIAL653195-25G
SIAL653195-5G
(2-Brom-4-methoxyphenyl)essigsäure, Sigma-Aldrich®
(2-Brom-4-methoxyphenyl)essigsäure
Formel:
C₉H₉BrO₃ Molecular Weight: 245,07 g/mol |
MDL:
MFCD02664700 CAS-Nummer: 66916-99-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...