3,5-Dimethoxyphenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3,5-Dimethoxybenzeneboronic acid

715271-1G 715271-5G
SIAL715271-1GEA 33.6 EUR
SIAL715271-1G SIAL715271-5G
3,5-Dimethoxyphenylboronsäure, Sigma-Aldrich®
3,5-Dimethoxyphenylboronsäure
Formel: C₈H₁₁BO₄
Molecular Weight: 181,98 g/mol
MDL: MFCD03095127
CAS-Nummer: 192182-54-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR