1-Chlor-5-(triethylsilyl)-4-pentin

Lieferant: CHEMPUR SILANE

SILS04790.1 SILS04790.5 SILS04790.25
SILNSILS04790.1EA 0 EUR
SILNSILS04790.1 SILNSILS04790.5 SILNSILS04790.25
1-Chlor-5-(triethylsilyl)-4-pentin
1-Chlor-5-(triethylsilyl)-4-pentin
Formel: C₁₁H₂₁ClSi
MDL: MFCD00671352
CAS-Nummer: 174125-30-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR