1-Chlor-5-(trimethylsilyl)-4-pentin

Lieferant: CHEMPUR SILANE

SILS04812.1 SILS04812.5 SILS04812.25
SILNSILS04812.1EA 0 EUR
SILNSILS04812.1 SILNSILS04812.5 SILNSILS04812.25
1-Chlor-5-(trimethylsilyl)-4-pentin
1-Chlor-5-(trimethylsilyl)-4-pentin
Formel: C₈H₁₅ClSi
Molecular Weight: 174.75 g/mol
MDL: MFCD00671353
CAS-Nummer: 77113-48-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR